Documents
Z247723
Coors high-alumina crucible cover
diam. 39.4 mm
About This Item
Code UNSPSC :
41122200
Nomenclature NACRES :
NB.43
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Coorstek 65608
Diamètre
39.4 mm
Description générale
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
For Z247626
Informations légales
Coors is a trademark of Coorstek, Inc.