Documents
Z215929
Solid PTFE stopper
joint: ST/NS 24/40
Synonyme(s) :
flask stopper, lab stopper, stoppers
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS24/40
Fabricant/nom de marque
Cowie 010.124
Description générale
Machined from solid PTFE with trapezoidal seals for medium to high vacuum performance up to 280°C (540°F).