Principaux documents
Z243868
Sigma-Aldrich
POWERMITE proportional on/off type-1,000W capacity heat control
EB-1C
1 EA
524,00 $
524,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
524,00 $
About This Item
Code UNSPSC :
41112303
Nomenclature NACRES :
NB.76
524,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Entrée CA/CC
120 V AC
Caractéristiques
CSA approved
Disponibilité
not available in EU
Poids
1.6 lb
Description générale
Model EB-1C has a cast aluminum case with gray, baked-on finish, satin SS face plate with flush-mounted receptacle and dial plate, pilot lamp (two lamps on model EB-1C), and a 3 ft, 3-wire power cord with U.S. plug.
Used to control THERMOWELL heating mantles or other resistive heating circuits (hot plates, heat tapes, water baths, etc.). Available in solid-state or proportional on/off models. All models are fused (without circuit breakers). Each unit is calibrated to deliver a known amount of power or voltage at a given dial setting in the 15 to 70% range (off below 15% and full on above 70%). High capacity with constant power output despite large line voltage fluctuations. Does not vary voltage. 50/60 Hz.
Informations légales
POWERMITE is a trademark of Laboratory Craftsmen, Inc.