Z218707
Clamps
fits flask, 2,000 ‑ 4,000 mL
Synonyme(s) :
flask clamp, flask lab, lab flask, reaction flasks
About This Item
Produits recommandés
Caractéristiques
fits flask 2,000 ‑ 4,000 mL
Fabricant/nom de marque
Ace Glass 650811
Description générale
Two-piece, black anodized aluminum clamp has three wing nuts to secure Ace reaction flask to head. Clamps also fit Pyrex reaction kettles.