Documents
Z174246
Sigma-Aldrich
Drying tower
joint: ST/NS 45/50
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.79
Joint
joint: ST/NS45/50
Description générale
Threaded ST/NS: 45/50 joint, glass stopcock with hose barb.
Liaison
See the Threaded Ground Glass Tutorial to see how threaded joints work.