Principaux documents
Z418137
Sigma-Aldrich
Aldrich® System 45 powder addition funnel
capacity 50 mL
Synonyme(s) :
funnel powder, powder funnel
1 EA
580,00 $
580,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
580,00 $
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
580,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Taille de la vis sans fin
small
Joint
joint ST/NS14/20
Hauteur
~180 mm
Capacité
50 mL
Catégories apparentées
Description générale
These funnels are designed to deliver free-flowing granular material to reactions by turning the auger feed. Delivery can be continuous or a single charge. Funnels are designed with large top openings with GL 45 threaded closures. PTFE auger can be removed easily for cleaning and is designed for vacuum applications. Only the 500 mL size has a clean out side-arm port. Supplied complete with auger and top cap.
Autres remarques
When assembling the auger into the remaining equipment, the auger unit must be fully advanced past the glass threads. Use water to moisten the o-ring when necessary. Use proper personal protective equipment (PPE) when handling glassware.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak
Pièce de rechange