Z169013
Porcelain berl saddles
pkg of 250 ea
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Produits recommandés
Conditionnement
pkg of 250 ea
Description générale
6 mm. Ideal for vacuum distillation, absorption, and extraction. Surface area: 1.15 m2/m3 In packages of approx. 250 (200 cc).
Maximum temperature: 1100°C.
Maximum temperature: 1100°C.