Principaux documents
Z156809
Sigma-Aldrich
Justrite® Type 1 safety can
capacity 1 gal
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
Matériaux
terneplated steel
Fabricant/nom de marque
Justrite 10301
H × D.E.
11 1/2 in. × 7 1/4 in.
Capacité
1 gal
Description générale
For the safe transport and storage of gasoline and other flammable liquids. All cans have a double brass, wire-mesh flame arrester and a self-closing spout for automatic pressure-relief. Not for use with corrosive solvents or solvents with water. Red enamel finish. FM approved and UL listed. Meets OSHA requirements.
Informations légales
Justrite is a registered trademark of Justrite Manufacturing Co.