Principaux documents
TEM-FF150CU50
Sigma-Aldrich
Formvar Supported Copper Square Mesh
size 150 mesh, box of 50
Synonyme(s) :
FORMVAR FILM 150 MESH VU 50/ BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
150 mesh
Description générale
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 150 mesh. All of our film is laid on the shiny side of the grid.