Principaux documents
TEM-CF400NI50
Sigma-Aldrich
Carbon Film Supported Nickel Square Mesh
size 400 mesh, box of 50
Synonyme(s) :
CARBON FILM 400 MESH NI 50/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
400 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Nickel grid, 400 mesh.