Documents
TEM-CF300AUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Gold Square Mesh
size 300 mesh, box of 50
Synonyme(s) :
CARBON SQUARE - MESH - AU 300 MESH UL 50/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
300 mesh
Description générale
A 3-4 nm ultrathin film of pure carbon supported standard squre mesh grids. Gold grid, 300 mesh.