Principaux documents
TEM-CF100HCU
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 100 mesh, box of 25
Synonyme(s) :
CARBON ONLY ON HEX COPPER GRIDS 25/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Matériaux
carbon film
copper grid
Description
film layer thickness 5 - 6 nm carbon
Conditionnement
box of 25
Taille
100 mesh
Description générale
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 100 mesh. Carbon film show better electrical and heat conduction in the electron beam.