Principaux documents
TEM-76042-41
Sigma-Aldrich
Micronporous Silicon Nitride TEM Window Grids for Cryo-EM Suspension
50nm Thick, pack of 10
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le produit TEM-76042-41 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Conditionnement
pack of 10
Description générale
Silicon nitrode microporous TEM window with single square of 500x500 micron size, and 50 nm thick SiN. Plasma cleanable, less than 0.5nm variation in film thickness, tolerate temperatures above 1000 ˚C, provides flat, insulating and hydrophobic surfaces