Yes, graphene generally should be coated by PMMA before transferred to the grid. And PMMA could be removed after transfer.
931152
TEM Grids
Holey Carbon Supported Copper Grids, grid size 300 mesh, hole avg. diam. 100 nm, box of 25
Synonyme(s) :
TEM grids
312,00 $
About This Item
312,00 $
Produits recommandés
Nom du produit
Holey Carbon Supported Copper Grids, grid size 300 mesh, hole avg. diam. 100 nm, box of 25
Matériaux
carbon film (holey)
copper grid
Conditionnement
box of 25
Taille de la grille
300 mesh
hole avg. diam.
100 nm
-
I want to know whether the graphene is coated by PMMA or not before transfered to the grid.