Principaux documents
SYNS531018
Synthware PTFE stopper with extraction nut
joint: ST/NS 10/18
Synonyme(s) :
Synthware S531018
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS10/18
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S531018
Description générale
PTFE stopper has a Nylon extraction nut permitting easy removal of the stopper from the glass outer joint.
Informations légales
Synthware is a trademark of Kemtech America Inc.