Documents
Z268798
Safe-Lab Thermometer/Tubing Adapter
joint: ST/NS 14/20, opening size 6-7 mm
Synonyme(s) :
thermometer holder
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS14/20
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F20309-1680
Taille de l'ouverture
6-7 mm
Description générale
PTFE body with Viton® O-ring and PP screw-cap with inside Viton O-ring for air-tight work. Loosen cap for depth adjustment of thermometer or tubing.
Informations légales
Safe-Lab is a trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company