Documents
SYNS191125
Synthware metering valve stopcock plug
bore size 2 mm, plug size: 11/25
Synonyme(s) :
Synthware S191125
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S191125
Taille de l'alésage
2 mm
Description générale
With a threaded PTFE needle valve for the fine control of flow through the bore of the stopcock plug. This replacement kit is complete with all the components.
Informations légales
Synthware is a trademark of Kemtech America Inc.