SYNF419950
Synthware™ two neck round bottom flask with angled side neck
50 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22
Synonyme(s) :
Synthware F419950, 2-neck round flask
About This Item
Produits recommandés
Matériaux
borosilicate glass flask
round bottom
two-neck (angled side neck)
Joint
center joint: ST/NS19/22
side joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F419950
Capacité
50 mL
50 mL
Catégories apparentées
1 of 4
Cet article | SYNF414450 | SYNF411150 | SYNF414150 |
---|---|---|---|
capacity 50 mL , 50 mL | capacity 50 mL , 50 mL | capacity 50 mL , 50 mL | capacity 50 mL , 50 mL |
material borosilicate glass flask, two-neck (angled side neck), round bottom | material borosilicate glass flask, round bottom , two-neck (angled side neck) | material borosilicate glass flask, round bottom , two-neck (angled side neck) | material borosilicate glass flask, round bottom , two-neck (angled side neck) |
joint center joint: ST/NS19/22, side joint: ST/NS19/22 | joint center joint: ST/NS24/40, side joint: ST/NS24/40 | joint center joint: ST/NS14/20, side joint: ST/NS14/20 | joint center joint: ST/NS24/40, side joint: ST/NS14/20 |
packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |