Documents
SYNF150025
Synthware micro filter flask
stopper size #3, capacity 25 mL
Synonyme(s) :
Synthware F150025
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
flat bottom flask
glass flask
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F150025
Capacité
25 mL
Description générale
Top is tooled for a #2 neoprene filter adapter or #3 rubber stopper.
Informations légales
Synthware is a trademark of Kemtech America Inc.