Principaux documents
Z254231
Sigma-Aldrich
Filter Adapter Set
seven black neoprene rubber adapters, sizes 1 through 7
Synonyme(s) :
Adapter Set, Filter Adapter Kit
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
Matériaux
neoprene
rubber
Fabricant/nom de marque
Aldrich
Description générale
Set of seven black neoprene rubber adapters for Buchner, conical, fritted-glass, and porcelain filter funnels. One of each size 1 through 7. Withstands autoclaving.
Les clients ont également consulté
Slide 1 of 3
1 of 3