Principaux documents
SYND351403
Synthware cow type graduated distillation receiver
joint: ST/NS 14/20
Synonyme(s) :
Synthware D351403, cow type distilling receiver
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Fabricant/nom de marque
D351403
Catégories apparentées
Description générale
Synthware distillation cow receiver features a standard taper joint (14/20) and four tubes graduated from 0-3mL in 0.2mL subdivisions.
Informations légales
Synthware is a trademark of Kemtech America Inc.