Principaux documents
SYNA201024
Synthware reducing adapter
female joint: ST/NS 10/30, male joint: ST/NS 24/40
Synonyme(s) :
Synthware A201024, glass reducing adapter, connecting adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
female joint: ST/NS10/30
male joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A201024
Description générale
Synthware reducing adapter is used for connecting dissimilar standard taper joint sizes. The bottom joint (24/40) is larger than the top joint (10/30).
Informations légales
Synthware is a trademark of Kemtech America Inc.