Principaux documents
SLW3152/08A
Pyrex® Volumetric flask, heavy duty, class A, amber, USP/ISO/DIN tolerances,
capacity 25 mL
Synonyme(s) :
Pyrex, SciLabware 3152/08A
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Le produit SLW3152/08A n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
borosilicate glass flask
Joint
joint ST/NS neck10/19
Caractéristiques
accuracy class A (ISO 1042 and DIN 12664, United States Pharmacopoeia (USP),conformity ′H′ certified)
Conditionnement
pkg of 5 ea
Fabricant/nom de marque
SciLabware 3152/08A
H totale
110 mm
Capacité
25 mL
Description générale
  • Complies with ISO 1042 and DIN 12664
  • Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
  • Manufactured from borosilicate glass with an amber coating on exterior surfaces to facilitate use with light sensitive solutions
  • Each flask is laser etched with an individual serial number, batch code and date of manufacturer
Informations légales
PYREX is a registered trademark of Corning, Inc.