Principaux documents
M9775
Sigma-Aldrich
4-(N-Maleimido)benzophenone
Synonyme(s) :
Benzophenone-4-maleimide
About This Item
Formule empirique (notation de Hill) :
C17H11NO3
Numéro CAS:
Poids moléculaire :
277.27
Numéro MDL:
Code UNSPSC :
12352100
ID de substance PubChem :
Nomenclature NACRES :
NA.22
Pertinence de la réaction
reagent type: cross-linking reagent
Solubilité
chloroform: 50 mg/mL
DMF: soluble
Température de stockage
2-8°C
Chaîne SMILES 
O=C1C=CC(=O)N1c2ccc(cc2)C(=O)c3ccccc3
InChI
1S/C17H11NO3/c19-15-10-11-16(20)18(15)14-8-6-13(7-9-14)17(21)12-4-2-1-3-5-12/h1-11H
Clé InChI
OZIZEXQRIOURIJ-UHFFFAOYSA-N
Application
A heterobifunctional cross-linking reagent containing a sulfhydryl-specific group and a photo-active group. Typically, coupled initially by thioether to molecule containing free sulfhydryl buffered at pH 6.8 (6.5-7.0). Second bonding occurs during UV irradiation (250 nm) via diradical excited state. Benzophenones demonstrate greater specificity for C-H insertion and are more stable in water than analogous reagents. In general they are more efficient in attachment because they may be repeatedly irradiated; however, a more intense irradiation may be required. The benzophenone is not sensitive to reduction compared to analogous reagents.
Pictogrammes
Exclamation mark
Mention d'avertissement
Warning
Mentions de danger
Classification des risques
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Organes cibles
Respiratory system
Code de la classe de stockage
11 - Combustible Solids
Classe de danger pour l'eau (WGK)
WGK 3
Point d'éclair (°F)
Not applicable
Point d'éclair (°C)
Not applicable
Équipement de protection individuelle
dust mask type N95 (US), Eyeshields, Gloves
Y Doi et al.
FEBS letters, 301(1), 99-102 (1992-04-13)
It has been shown that the EGTA-resistant actin, one of the two actin molecules associated to gelsolin, can be predominantly cross-linked to gelsolin by benzophenone-4-maleimide (BPM), a photoaffinity-labeling reagent, which was conjugated to Cys-374 of actin prior to cross-linking (Doi
J F Clémence et al.
Bioconjugate chemistry, 6(4), 411-417 (1995-07-01)
To attain light-dependent functionalization of biocompatible materials, a photolabel-derivatized, bioactive laminin fragment has been synthesized, chemically characterized, and photoimmobilized. Covalent high-resolution patterning of the laminin fragment CDPGYIGSR to hydroxylated fluorinated ethylene propylene (FEP-OH), poly(vinyl alcohol), and glycophase glass has been
Takao Inoue et al.
The Journal of biological chemistry, 280(30), 27896-27903 (2005-06-14)
The vacuolar (H+)-ATPases (V-ATPases) are multisubunit complexes responsible for ATP-dependent proton transport across both intracellular and plasma membranes. The V-ATPases are composed of a peripheral domain (V1) that hydrolyzes ATP and an integral domain (V0) that conducts protons. Dissociation of
Mayumi Tamura et al.
Biological & pharmaceutical bulletin, 34(6), 929-932 (2011-06-02)
LEC-1 is the first tandem repeat-type galectin isolated from an animal system; this galectin has two carbohydrate recognition domains in a single polypeptide chain. Because its two lectin domains have different sugar-binding profiles, these domains are thought to interact with
L Sahlman et al.
The Journal of biological chemistry, 272(47), 29518-29526 (1997-12-31)
Bacterial detoxification of mercuric ion depends on the presence of one or more integral membrane proteins (MerT and/or MerC) whose postulated function is in transport of Hg2+ from a periplasmic Hg2+-binding protein (MerP) to cytoplasmic mercuric reductase. In this study