HSWNH161
HSW FINE-JECT hypodermic needles, disposable
gauge 16, L 1 in., white, sterile
About This Item
Code UNSPSC :
42142615
Nomenclature NACRES :
NB.03
Produits recommandés
Matériaux
white
Description
thin wall
Stérilité
sterile
Fabricant/nom de marque
Henke Sass Wolfe
Longueur
1 in.
Calibre
16
Description générale
- Disposable, sterile, FDA registered
- ISO 9626/1991 stainless steel
- Color-coded polypropylene hub
- Protective polypropylene cap
- Siliconized needle tube
- Pyrogen-free and non-toxic
- 5-year shelf life