Principaux documents
BR36981
BRAND® volumetric flask BLAUBRAND® USP conformity certified
capacity 250 mL, neck joint: ST/NS 14/23, glass stopper
Synonyme(s) :
BLAUBRAND®, USP, Volumetric flasks
2 EA
152,00 $
152,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
2 EA
152,00 $
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
152,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass
glass stopper
Joint
joint: ST/NS neck14/23
Caractéristiques
accuracy class A
Conditionnement
pack of 2
Fabricant/nom de marque
BRAND 36981
Capacité
250 mL
Catégories apparentées
Description générale
The BRAND® volumetric flask BLAUBRAND® USP conformity certified is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for a variety of laboratory applications. Each flask comes with a USP batch certificate, confirming its adherence to stringent quality and safety standards. This volumetric flask is an essential tool for precise analytical work, making it ideal for laboratories that require reliable and certified liquid handling solutions.
Informations légales
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG