Principaux documents
BR150070
BRAND® watch glass
soda-lime glass, diam. 150 mm , ground edges stress-reduced
Synonyme(s) :
Dishes, Watch glasses
10 EA
120,00 $
120,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
10 EA
120,00 $
About This Item
Code UNSPSC :
41121801
Nomenclature NACRES :
NB.43
120,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
soda-lime glass
Fabricant/nom de marque
BRAND 150070
Diamètre
150 mm , ground edges stress-reduced
Description générale
The BRAND® watch glass is made from soda-lime glass and complies with DIN 12341 standards. It features ground edges for safe handling and a refined finish. This watch glass is stress-reduced to minimize the risk of shattering, enhancing safety during laboratory use. Ideal for covering beakers or evaporating dishes, it provides an effective solution for preventing contamination and evaporation of samples. Its durable design makes it a reliable tool in various laboratory applications, ensuring optimal performance in your experiments.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG