Principaux documents
BR148030
BRAND® PP powder funnel short wide stem
top diam. 100 mm, bottom diam. 25 mm
Synonyme(s) :
Funnels, Powder funnels
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Conditionnement
pack of 20 ea
Fabricant/nom de marque
BRAND 148030
Diam. du fond
25 mm
Diam. de la partie supérieure
100 mm
Caractéristiques et avantages
  • BRAND® powder funnel is ideal for transfering powders.
  • Made from break-resistant polypropylene, these general lab use funnels include a short wide stem that facilitates optimal filtration.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG