Principaux documents
BR146519
BRAND® glass powder funnel short wide stem
top diam. 100 mm, bottom diam. 25 mm
Synonyme(s) :
Funnels, Power funnels
1 EA
26,80 $
26,80 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
26,80 $
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
26,80 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 146519
Diam. du fond
25 mm
Diam. de la partie supérieure
100 mm
Description générale
The BRAND® Glass Powder Funnel with Short Wide Stem is expertly crafted from Boro 3.3 glass, ensuring durability and high performance in laboratory applications. This funnel features a ground and fire-polished upper rim, providing a smooth edge for safe handling and easy pouring. Designed with a short, wide stem, it facilitates efficient transfer of powders and granular materials, minimizing the risk of clogs and ensuring a swift flow.
Caractéristiques et avantages
  • BRAND® borosilicate glass powder funnel is best suited for transferring powders.
  • These general lab use funnels include ground and fire-polished upper rim.
  • Features a short, wide stem that facilitates optimal filtration.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG