Principaux documents
BR114823
BRAND® PP graduated centrifuge tube, screw cap
volume 50 mL, with base, sterile
Synonyme(s) :
Centrifuge tubes, Tubes
250 EA
391,00 $
391,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
250 EA
391,00 $
About This Item
Code UNSPSC :
41121703
Nomenclature NACRES :
NB.11
391,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polypropylene
screw top cap
translucent tube
Stérilité
sterile
Caractéristiques
RCF 3,000 × g
with base
Conditionnement
pack of 250
Fabricant/nom de marque
BRAND 114823
Volume
50 mL
Description générale
The BRAND® PP Graduated Centrifuge Tube with Screw Cap is made from high-quality polypropylene (PP) and features a translucent design for easy visibility of contents. This tube is engineered to withstand relative centrifugal forces (RCF) of up to 3000, making it suitable for various laboratory applications. It comes graduated, allowing for precise measurements, and is available in both non-sterile and gamma-ray sterilized options in 15 ml (graduated up to 13 ml) or 50 ml capacities. The screw cap, made from polyethylene (PE) and equipped with a sealing cone, ensures a secure closure to prevent leaks. This centrifuge tube offers high clarity and reliability, making it an essential tool for researchers and technicians in the lab.
Caractéristiques et avantages
  • BRAND® graduated centrifuge tube is the ideal solution for all your centrifuge needs.
  • Made from highly transparent polypropylene (PP) for enhanced sample visibility.
  • Bio-inert PP offers optimal cell and protein recovery.
  • Includes a screw cap with a plug for preventing sample loss.
  • Tubes are tested to withstand RCF up to 3000.
  • γ-ray sterilized.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG