Principaux documents
BAH379350000
SP Bel-Art® FEP Coated Steel Forceps
straight PTFE-coated stainless steel forceps, L 98 mm , tip coating 38mm
Synonyme(s) :
stainless steel forceps
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Matériaux
straight PTFE-coated stainless steel forceps
Conditionnement
pkg of 2 ea (48/Case)
Fabricant/nom de marque
Bel-Art H37935-0000
Longueur
98 mm , tip coating 38mm
Description générale
The Brown PTFE FEP coating on these stainless steel forceps will not scratch or mar delicate surfaces, while its anti-stick qualities prevent material build-up.
Caractéristiques et avantages
  • Coating complies with FDA regulations for direct contact with food
  • All forceps are 9.8cm (31/2") long with medium points and serrated tips
  • Select the coated length ideal for your use
  • PTFE is a registered trademark of The Chemours Company FC LLC and used under license by Bel-Art Products, Inc.
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP