MilliporeSigma
Toutes les photos(8)
Principaux documents
BAF371090020
SP Bel-Art® Spinbar® magnetic stir bar
octagonal yellow PTFE-coated, L × diam. 1.5 in. (38.1 mm) × 0.3125 in. (8 mm)
Synonyme(s) :
Spinbar® magnetic stir bar
1 EACH
18,70 $
18,70 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EACH
18,70 $
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.51
18,70 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
octagonal yellow PTFE-coated
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F37109-0020
L × diam.
1.5 in. (38.1 mm) × 0.3125 in. (8 mm)
Description générale
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
Leakproof, seamless, polished, and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape. Choose from red, yellow or blue.
Caractéristiques et avantages
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE brand PTFE Coating – Superior Chemical Compatibility
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP