Documents
756393
Sigma-Aldrich
Aluminum nitride
powder, -200 mesh, 99.8% trace metals basis
Synonyme(s) :
Aluminum mononitride
About This Item
Formule linéaire :
AlN
Numéro CAS:
Poids moléculaire :
40.99
Numéro CE :
Numéro MDL:
Code UNSPSC :
12352300
ID de substance PubChem :
Nomenclature NACRES :
NA.23
Pureté
99.8% trace metals basis
Forme
powder
Taille des particules
-200 mesh
Pf
>2200 °C (lit.)
Densité
3.26 g/mL at 25 °C (lit.)
Chaîne SMILES 
N#[Al]
InChI
1S/Al.N
Clé InChI
PIGFYZPCRLYGLF-UHFFFAOYSA-N
Application
Aluminum nitride is a refractory ceramic that is used as an electrically insulating material in applications similar to aluminum oxide. AlN is also used in optical and semiconductor devices with GaN to produce LEDs on sapphire and in piezoelectric MEMS devices.
Pictogrammes
Health hazardEnvironment
Mention d'avertissement
Danger
Mentions de danger
Classification des risques
Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1 Inhalation
Organes cibles
Lungs
Code de la classe de stockage
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Classe de danger pour l'eau (WGK)
WGK 3
Point d'éclair (°F)
Not applicable
Point d'éclair (°C)
Not applicable
Les clients ont également consulté
Slide 1 of 1
1 of 1
Ambacher, O.
Journal of Physics D: Applied Physics, 31, 2653-2653 (1998)
Fang, X.; et al.
Journal of Materials Chemistry, 18, 509-509 (2008)
Strite, S.; Morkoc, H.
J. Vac. Sci. Technol. B, 10, 1237-1237 (1992)
Yong-Bing Tang et al.
ACS nano, 5(5), 3591-3598 (2011-04-13)
Arrays of well-aligned AlN nanowires (NWs) with tunable p-type conductivity were synthesized on Si(111) substrates using bis(cyclopentadienyl)magnesium (Cp(2)Mg) vapor as a doping source by chemical vapor deposition. The Mg-doped AlN NWs are single-crystalline and grow along the [001] direction. Gate-voltage-dependent
Sangbok Park et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2012, 5683-5686 (2013-02-01)
In this paper, we present an antenna system for microwave non-invasive hyperthermia lipolysis. The antenna system consists of a circular waveguide antenna radiating electromagnetic waves, AlN (Aluminum Nitride) radome and heat sink. The AlN radome with heat sink helps to
Articles
Three approaches generate white light, including LED-based down-conversion for broader applications.