MilliporeSigma
TLC Analysys of Antioxidative Flavonoids in Plant Extracts on Silica Gel 60 F254s
TLC Analysys of Antioxidative Flavonoids in Plant Extracts on Silica Gel 60 F254s application for TLC
Materials
accessory
Product No.
Description
Pricing
Bottle-type sprayer
capacity 240 mL
Rectangular TLC developing tanks, complete
L × H × W 27.0 cm × 26.5 cm × 7.0 cm
Sigma TLC spray box
Spectroline® E-Series UV lamp
output8 W, AC/DC input 115 V AC
Rectangular TLC developing tanks, complete
L × H × W 7.5 cm × 15.5 cm × 8.0 cm
Hamilton® TLC syringes
1702, volume 25 μL, needle size 22s ga, needle L 51 mm (2 in.),
coated tip
Spectroline® E-Series UV lamp
output6 W, AC/DC input 120 V AC
Hamilton® TLC syringes
1705, volume 50 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Hamilton® TLC syringes
1701, volume 10 μL, needle size 26s ga, needle type, fixed, needle L 51 mm (2 in.),
coated tip
Rectangular TLC developing tanks, complete
L × H × W 17.5 cm × 16.0 cm × 8.2 cm
Spectroline® E-Series UV lamp
output6 W, AC/DC input 230 V AC
Spectroline® E-Series UV lamp
output4 W, AC/DC input 230 V AC
Spectroline® E-Series UV lamp
output8 W, AC/DC input 230 V AC
Latch-lid TLC developing chambers
For use with 10 cm × 10 cm plates
Hamilton® TLC syringes
1710, volume 100 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Latch-lid TLC developing chambers
For use with 20 cm × 20 cm plates
Spectroline® E-Series UV lamp
output4 W, AC/DC input 115 V AC
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
capacity 20 μL, accuracy: ±0.5%
Reagent sprayer for TLC plates
volume 250 mL, Bottle-Type
CONDITIONS
sample preparation
Extract samples were filtered with a 0.45 μm syringe filter. Application by CAMAG ATS4 6mm bandwise.
plate
1.15445 HPTLC LiChrospher Silica gel 60 F254s 20 cm x 10 cm (1.15445.0001)
eluent
[A] ethylacetate; [B] formic acid; [C] glacial acid; [D] water; (100:11:11:27, A:B:C:D) containing 0.1% n-Heptane
migration distance
5 cm
chamber
normal chamber without chamber saturation
detector
366 nm Diphenylboric acid-2-aminoethylester-reagent for Flavonoids, Vanillin-sulfuric acidreagent e.g. for Phenols, Steroids, Glycoside; DPPH reagent for antioxidants (visible light)