Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z266019

Supelco

Latch-lid TLC developing chambers

For use with 10 cm × 10 cm plates

Synonym(s):

Locking Lid Thin-layer chromatography developing chambers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

technique(s)

thin layer chromatography (TLC): suitable

Looking for similar products? Visit Product Comparison Guide

General description

Latch-Lid units have a unique stainless steel latching device that holds the lid and tank firmly in place. This permits complete saturation of the chamber with solvent vapor for uniform development of chromatograms. Glass tank features flat ground tops, inner and outer rims beveled for safety, and flat ground bottoms for stability. Accessorize with multi-plate TLC racks for simultaneous development of up to six TLC plates.

Racks not included.

Application



  • Toxicity Assessment of Nickel Using Aspergillus niger and Its Removal from an Industrial Effluent.: This study focuses on determining the Nickle concentration using Aspergillus niger and explores its removal from industrial effluent. The research provides valuable analytical techniques for environmental chemists focusing on bioremediation and industrial waste management (Rajendran et al., 2002).




Legal Information

Latch-Lid is a trademark of General Glassblowing, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Srishti Dar et al.
Nature protocols, 12(2), 390-400 (2017-01-27)
The process of membrane fission is fundamental to diverse cellular processes such as nutrient uptake, synaptic transmission and organelle biogenesis, and it involves the localized application of curvature stress to a tubular membrane intermediate, forcing it to undergo scission. Alternative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service