Skip to Content
MilliporeSigma
All Photos(2)

Documents

Z264415

Supelco

Hamilton® TLC syringes

1710, volume 100 μL, needle size 22 ga, needle L 51 mm (2 in.),

coated tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

stainless steel needle (PTFE coated tip)

needle size

22 ga

feature

needle type fixed

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 81050

needle L

51 mm (2 in.)

volume

100 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hamilton syringe is available in gas-tight type that minimizes the gas exchange in analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.
PTFE coats the final 3/4 in. of this 2 in. (51 mm) long cemented needle. This allows for more reproducible sample spotting on TLC plates.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques
Don P. Wolf
Springer Science & Business Media, 192-192 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service