Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58685

Flangeless Fitting

1/4-28 male Upchurch nut and ferrule for 1/16 in. O.D. tubing

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$56.50

$56.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 EA
$56.50

About This Item

UNSPSC Code:
41121815

$56.50


Please contact Customer Service for Availability

Request a Bulk Order

material

Delrin® nut
Tefzel® ferrule (flangeless)

packaging

pkg of 5 ea

parameter

1000 psi max. pressure (70 bar)

fitting

1/4-28 male Upchurch nut and ferrule for 1/16 in. O.D. tubing

Legal Information

Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service