Key Documents
56701
Upchurch Flangeless Ferrule
1/4-28 UNF ferrule for 1/8 in. O.D. tubing (requires Flangeless nut), ferrule only
Synonym(s):
Ferrules, 1/8in, Tefzel®, 10 pk
Select a Size
10 PKG
$33.70
$33.70
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
10 PKG
$33.70
About This Item
UNSPSC Code:
41115700
$33.70
Please contact Customer Service for Availability
Request a Bulk Order
material
Tefzel® ferrule
packaging
pkg of 10 ea
manufacturer/tradename
Upchurch Scientific, Inc P-300X
fitting
1/4-28 UNF ferrule for 1/8 in. O.D. tubing (requires Flangeless nut)
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 3
1 of 3