Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48556

Supelco

Phenol

analytical standard

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

autoignition temp.

1319 °F

expl. lim.

8.6 %

packaging

vial of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

application(s)

environmental

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhi-Ling Guo et al.
Brain research, 1229, 125-136 (2008-07-29)
The midline medulla oblongata, which includes the nucleus raphe obscurus, raphe magnus and raphe pallidus (NRP), is involved in regulation of cardiovascular responses. Opioids and serotonin (5-HT) are thought to function as important neurotransmitters in this region. We previously have
Shinya Nagashima et al.
Bioorganic & medicinal chemistry, 16(13), 6509-6521 (2008-06-07)
Signal transducers and activators of transcription 6 (STAT6) is a key regulator of the type 2 helper T (Th2) cell immune response and a potential therapeutic target for allergic diseases such as asthma and atopic diseases. To search for potent
Rohan A Davis et al.
Journal of medicinal chemistry, 54(6), 1682-1692 (2011-02-22)
In order to discover novel probes that may help in the investigation and control of infectious diseases through a new mechanism of action, we have evaluated a library of phenol-based natural products (NPs) for enzyme inhibition against four recently characterized
Marie A Colucci et al.
Journal of medicinal chemistry, 50(23), 5780-5789 (2007-10-20)
NAD(P)H:quinone oxidoreductase 1 is a proposed target in pancreatic cancer. We describe the synthesis of a series of indolequinones, based on the 5- and 6-methoxy-1,2-dimethylindole-4,7-dione chromophores with a range of phenolic leaving groups at the (indol-3-yl)methyl position. The ability of
Alessio Innocenti et al.
Bioorganic & medicinal chemistry, 18(6), 2159-2164 (2010-02-27)
A series of phenolic acids and phenol natural products, such as p-hydroxybenzoic acid, p-coumaric acid, caffeic acid, ferulic acid, gallic acid, syringic acid, quercetin, and ellagic acid, were investigated for their inhibitory effects against the metalloenzyme carbonic anhydrase (CA, EC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service