MilliporeSigma
  • Identification of 4-benzylamino-2-[(4-morpholin-4-ylphenyl)amino]pyrimidine-5-carboxamide derivatives as potent and orally bioavailable STAT6 inhibitors.
Identification of 4-benzylamino-2-[(4-morpholin-4-ylphenyl)amino]pyrimidine-5-carboxamide derivatives as potent and orally bioavailable STAT6 inhibitors.
Bioorganic & medicinal chemistry (2008-06-07)
Shinya Nagashima, Hiroshi Nagata, Masahiro Iwata, Masaki Yokota, Hiroyuki Moritomo, Masaya Orita, Sadao Kuromitsu, Akiko Koakutsu, Keiko Ohga, Makoto Takeuchi, Mitsuaki Ohta, Shin-ichi Tsukamoto
ABSTRACT
Signal transducers and activators of transcription 6 (STAT6) is a key regulator of the type 2 helper T (Th2) cell immune response and a potential therapeutic target for allergic diseases such as asthma and atopic diseases. To search for potent and orally bioavailable STAT6 inhibitors, we synthesized a series of 4-benzylaminopyrimidine-5-carboxamide derivatives and evaluated their STAT6 inhibitory activities. Among these compounds, 2-[(4-morpholin-4-ylphenyl)amino]-4-[(2,3,6-trifluorobenzyl)amino]pyrimidine-5-carboxamide (25y, YM-341619, AS1617612) showed potent STAT6 inhibition with an IC(50) of 0.70nM, and also inhibited Th2 differentiation in mouse spleen T cells induced by interleukin (IL)-4 with an IC(50) of 0.28 nM without affecting type 1 helper T (Th1) cell differentiation induced by IL-12. In addition, compound 25y showed an oral bioavailability of 25% in mouse.
Sigma-Aldrich
2,6-Dichlorophenol, 99%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
2-Chlorophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
2-Chlorophenol, ≥99%
Sigma-Aldrich
2-Chlorophenol, 98%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent
Sigma-Aldrich
Phenol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, TE-saturated, ~73% (T)