Key Documents
Z683191
Small Rodent Shipping Container
About This Item
UNSPSC Code:
41102601
NACRES:
NB.22
L × W × D
17.75 in. × 9 in. × 6 in., 159 total square inches. Configurable up to 2 compartments.
Application
Filtration Media:
  • Spunbond Polypropylene - 170 cfm/ft
Autoclave Cycles:
  • 2 cycles maximum recommended up to 126 ºC, 20 minutes maximum
  • Meets IATA (International Air Transport Association) specifications
  • All-plastic construction: box, lid, dividers and filter medium
  • Superior airflow via patented engineered openings
  • Easily stackable for material handling and transportation, with integral spacers to ensure proper air circulation
  • Clear plastic windows enables at-a-glance inspection of lab animals
  • Positive open/close lid facilitates animal insertion/removal
  • Interior rim provides a barrier against gang and go incidents
  • Manufactured in the USA
Chemical Resistance - Inert to most chemicals except:
  • 9Chloroform
  • Clorosulfonic Acid
  • Chlorine Gas
  • Carbon Tetrachloride
  • Ixooctane
  • Xylene
  • Nitric Acid
  • Petroleum Ether
  • Amyl chloride
  • Butyl Acetate
  • Benzene
  • Bromine Water
  • Most Petroleum fuels will attack
Features and Benefits
Premium Shipping Containers for Premium Research Models
  • Sigma-Aldrich offers plastic containers for shipping your research models more safely. These containers are sturdier than cardboard boxes and have excellent protection against contaminants, breakage, and leakage en route to your lab
  • The patented design also ensures unsurpassed convenience and ease of handling at every stage
Kit Components Only
Product No.
Description
  • Container 1 ea
  • Lid 1 ea
  • Large Closure Label 1 ea
  • Small Closure Label 2 ea
  • Live Animal Label 2 ea
Kit Components Also Available Separately
Product No.
Description
SDS
  • Small Rodent Box Divider 1 ea
Choose from one of the most recent versions:
Certificates of Analysis (COA)