Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z177814

Scienceware® polypropylene trays

L × W 30 cm × 20 cm

Synonym(s):

laboratory tray, plastic tray

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.51

material

polypropylene container

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art H16260-0000

L × W

30 cm × 20 cm

capacity

5 L (1.3 gal)

Looking for similar products? Visit Product Comparison Guide

General description

Lightweight molded polypropylene trays have rounded corners and smooth, tapered sides for nesting.

  • Chemical resistant and corrosion proof
  • Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
  • Steam autoclavable at 121°C (250°F)
  • Cover not included

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service