Key Documents
Z381845
Sigma-Aldrich
Ultramicro pipette tips
Microcapillary, round tip, maximum volume 10 μL, non-sterile
Select a Size
1 PKG
$96.70
$96.70
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
$96.70
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
10 μL
sterility:
non-sterile
packaging:
pack of 200 ea (Bag)
manufacturer/tradename:
Sorenson 1511
$96.70
Please contact Customer Service for Availability
material
colorless
polypropylene
polypropylene
sterility
non-sterile
packaging
pack of 200 ea (Bag)
manufacturer/tradename
Sorenson 1511
maximum volume
10 μL
volume
10 μL
General description
Low capacity improves accuracy with small sample volumes by reducing dead space.
Application
For loading sequencing gels 0.6 mm or thicker.
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)