Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TGEN-CP

Sigma-Aldrich

Cas9 Recombinant protein from Streptococcus pyogenes

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Product TGEN-CP is not currently sold in your country. Contact Technical Service

form

lyophilized powder

Quality Level

packaging

pkg of 250 μg
pkg of 50 μg
pkg of 500 μg

shipped in

ambient

storage temp.

−70°C

General description

Recombinant Cas9 proteins (∼160 KD) are ready-to-use reagents for genome engineering experiments. When combined with target-specific guide RNAs, Cas9 proteins will form ready-to-use nucleases suitable for transfection of cell cultures and for the accelerated development of genetically-modified animals via one-cell embryo injection.

Features and Benefits

  • Highly Specific
  • Highly Active
  • Ready-to-inject/transfect

Packaging

The kit comes in three pack sizes:
  • TGEN-CP-50UG
  • TGEN-CP-250UG
  • TGEN-CP-500UG

Components

Each kit consists of:
  • one vial of Cas9 recombinant protein
  • one vial containing 1 mL of 1x dilution buffer
  • one vial containing 1mL of Nuclease free water with glycerol

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB.

Linkage

Use our CRISPR Selection Tool to order gRNA at sigma.com/crisprs

Know more about Cas9 Recombinant proteins

Physical form

Lyophilized Recombinant _Sp_Cas9 protein

Preparation Note

It is recommended that the lyophilized Cas9 protein be resuspended in the nuclease-free water containing 50% glycerol, that is provided in the kit, to get the desired concentration.
The solution should be allowed to incubate at room temperature (20−23 °C) for 10 minutes. Centrifuge immediately before use.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service