Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TRACRRNAMOD

Sigma-Aldrich

SygRNA Cas9 Synthetic Modified tracrRNA

Sign Into View Organizational & Contract Pricing

Select a Size

5 NMOL
$154.00

$154.00


Estimated to ship onFebruary 03, 2025Details


Request a Bulk Order

Select a Size

Change View
5 NMOL
$154.00

About This Item

UNSPSC Code:
12352208
NACRES:
NA.51

$154.00


Estimated to ship onFebruary 03, 2025Details


Request a Bulk Order

packaging

pkg of 1 vial

shipped in

ambient

storage temp.

−20°C

General description

The SygRNA modified synthetic RNA system recruits WT SpCas9, eSpCas9, dCas9, or Cas9D10A to specific genomic targets. 2′ O-methyl and phosphothioate modifications have been incorporated to enhance the stability and performance of the synthetic RNA. The modified tracrRNA (trans-activating crRNA) binds to a crRNA (CRISPR RNA) and the resulting crRNA-tracrRNA complex guides SpCas9 and SpCas9 variant proteins to crRNA-specified sites in the genome. SygRNA modified synthetic tracrRNA and crRNAs are optimized for use with a variety of Cas9 expressing cell lines, Cas9 proteins, or Cas9 mRNAs and are compatible with a delivery methods such as microinjection, lipofection, and electroporation.

Application

Functional Genomics/Target Validation/Transgenics

  • Creation of gene knockouts in multiple cell lines
  • Complete knockout of genes not amenable to RNAi
  • Creation of knock-in cell lines with promoters, fusion tags, or reporters integrated into endogenous genes
  • Generation of transgenic animals via embryo microinjection or electroporation

Features and Benefits

HPLC-purified SygRNA modified tracrRNA oligo is optimized for use with a target-specific crRNA or modified crRNA and a source of SpCas9 protein for genome editing applications.

Linkage

The SygRNA Cas9 tracrRNA oligo is used in conjunction with a target-specific crRNA and a source of Cas9 protein to form active RNP complexes.

Visit SigmaAldrich.com/CRISPRessentials to to review available Cas9 sources.

Order crRNAs at SigmaAldrich.com/CRISPR

Preparation Note

Resuspend SygRNA modified tracrRNA oligo in a RNAase-free Tris-containing buffer with a pH 8.0 or less.

Legal Information

SygRNA is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Combine guaranteed sgRNAs with our comprehensive range of CRISPR products and tools, including Cas9 and delivery reagents, for efficient genome modification with higher specificity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service