SYNS386150
Synthware™ long syringe needle
O.D. × L 0.6 mm × 150 mm , Bevel Tip, needle, female luer lock
Synonym(s):
Synthware S386150, stainless steel syringe needle, bevel tip needle
Select a Size
$14.30
Select a Size
About This Item
$14.30
Recommended Products
feature
needle female luer lock
packaging
pack of 1 ea
manufacturer/tradename
Kemtech S386150
O.D. × L
0.6 mm × 150 mm , Bevel Tip
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service