Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3349

Sigma-Aldrich

SGC8158N

≥98% (HPLC)

Synonym(s):

9-((3aR,4R,6S,6aS)-6-(((4-(((4’-Chloro-[1,1’-biphenyl]-3-yl)methyl)amino)butyl)thio)methyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)-9H-purin-6-amine, 9-((3aR,4R,6S,6aS)-6-((4-((4’-Chlorobiphenyl-3-yl)methylamino)butylthio)methyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)-9H-purin-6-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H35ClN6O3S
Molecular Weight:
595.16
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C(C2=CC(CNCCCCSC[C@@H]3[C@@H]4[C@H]([C@H](N5C6=C(C(N)=NC=N6)N=C5)O3)OC(C)(O4)C)=CC=C2)C=C1

InChI

1S/C30H35ClN6O3S/c1-30(2)39-25-23(38-29(26(25)40-30)37-18-36-24-27(32)34-17-35-28(24)37)16-41-13-4-3-12-33-15-19-6-5-7-21(14-19)20-8-10-22(31)11-9-20/h5-11,14,17-18,23,25-26,29,33H,3-4,12-13,15-16H2,1-2H3,(H2,32,34,35)/t23-,25-,26-,29-/m1/s1

InChI key

BTSGMZSEWQTVAJ-CTDWIVFPSA-N

Biochem/physiol Actions

Negative control probe for selective and cell-active PRMT7 inhibitor SGC8158
SGC8158N is a negative control probe for selective and cell-active PRMT7 inhibitor SGC8158.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena M Szewczyk et al.
Nature communications, 11(1), 2396-2396 (2020-05-16)
Protein arginine methyltransferases (PRMTs) regulate diverse biological processes and are increasingly being recognized for their potential as drug targets. Here we report the discovery of a potent, selective, and cell-active chemical probe for PRMT7. SGC3027 is a cell permeable prodrug

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service