MilliporeSigma
  • Pharmacological inhibition of PRMT7 links arginine monomethylation to the cellular stress response.
Pharmacological inhibition of PRMT7 links arginine monomethylation to the cellular stress response.
Nature communications (2020-05-16)
Magdalena M Szewczyk, Yoshinori Ishikawa, Shawna Organ, Nozomu Sakai, Fengling Li, Levon Halabelian, Suzanne Ackloo, Amber L Couzens, Mohammad Eram, David Dilworth, Hideto Fukushi, Rachel Harding, Carlo C Dela Seña, Tsukasa Sugo, Kozo Hayashi, David McLeod, Carlos Zepeda, Ahmed Aman, Maria Sánchez-Osuna, Eric Bonneil, Shinji Takagi, Rima Al-Awar, Mike Tyers, Stephane Richard, Masayuki Takizawa, Anne-Claude Gingras, Cheryl H Arrowsmith, Masoud Vedadi, Peter J Brown, Hiroshi Nara, Dalia Barsyte-Lovejoy
ABSTRACT
Protein arginine methyltransferases (PRMTs) regulate diverse biological processes and are increasingly being recognized for their potential as drug targets. Here we report the discovery of a potent, selective, and cell-active chemical probe for PRMT7. SGC3027 is a cell permeable prodrug, which in cells is converted to SGC8158, a potent, SAM-competitive PRMT7 inhibitor. Inhibition or knockout of cellular PRMT7 results in drastically reduced levels of arginine monomethylated HSP70 family stress-associated proteins. Structural and biochemical analyses reveal that PRMT7-driven in vitro methylation of HSP70 at R469 requires an ATP-bound, open conformation of HSP70. In cells, SGC3027 inhibits methylation of both constitutive and inducible forms of HSP70, and leads to decreased tolerance for perturbations of proteostasis including heat shock and proteasome inhibitors. These results demonstrate a role for PRMT7 and arginine methylation in stress response.
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Anti-dimethyl-BAF155 Antibody (Arg1064, asymetrically di-methylated), from rabbit, purified by affinity chromatography
Sigma-Aldrich
L-Lysine-13C6,15N2 hydrochloride, 99 atom % 13C, 99 atom % 15N, 95% (CP)
Sigma-Aldrich
L-Arginine-13C6,15N4 hydrochloride, 99 atom % 13C, 99 atom % 15N, 95% (CP)
Sigma-Aldrich
MISSION® esiRNA, targeting human PRMT7
Sigma-Aldrich
SGC8158N, ≥98% (HPLC)