Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4391

Sigma-Aldrich

Brij® L4

Synonym(s):

Polyethylene glycol dodecyl ether, Polyoxyethylene (4) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C20H42O5)n
CAS Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

refractive index

n20/D 1.451 (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCO

InChI

1S/C20H42O5/c1-2-3-4-5-6-7-8-9-10-11-13-22-15-17-24-19-20-25-18-16-23-14-12-21/h21H,2-20H2,1H3

InChI key

WPMWEFXCIYCJSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 235989-Sigma-Aldrich | Brij L4 average Mn ~362

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Koizumi et al.
AJR. American journal of roentgenology, 199(1), 200-207 (2012-06-27)
Balloon-occluded retrograde transvenous obliteration has been traditionally based on liquid sclerotherapy. However, overdose and systemic spillage of liquid sclerosant can cause severe complications, such as hemolysis, which lead to hemoglobinuria, allergy, acute respiratory distress syndrome, and other disorders. The purpose
Jacomina W van den Esschert et al.
Annals of surgery, 255(2), 311-318 (2012-01-14)
To compare the safety and hypertrophy response after portal vein embolization (PVE) using 2 absorbable and 3 permanent embolization materials. Portal vein embolization is used to increase future remnant liver volume preoperatively. Application of temporary, absorbable embolization materials could be
Otte Brosjö et al.
Acta orthopaedica, 84(5), 502-505 (2013-11-01)
Recent data suggest that percutaneous sclerotherapy is a safe alternative to surgery for treatment of aneurysmal bone cysts (ABCs). We present our experience of this method. We retrospectively analyzed data from 38 consecutive patients treated with repeated injections of polidocanol.
S Rathbun et al.
Phlebology, 27(3), 105-117 (2012-02-22)
Endovenous foam sclerotherapy (EFS) is used widely throughout the USA for the treatment of venous disorders. The purpose of the quantitative meta-analysis was to systematically and comprehensively evaluate the literature to provide accurate estimates of safety and efficacy outcomes for
Amish Patel et al.
AJR. American journal of roentgenology, 199(4), 721-729 (2012-09-22)
The purpose of this review is to describe the clinical factors related to balloon-occluded retrograde transvenous obliteration, including the preparation needed, the technique and challenges, and the outcomes. Although the procedure can be performed when transjugular intrahepatic portosystemic shunt is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service