Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

235989

Sigma-Aldrich

Brij® L4

average Mn ~362

Synonym(s):

Polyethylene glycol dodecyl ether, Polyoxyethylene (4) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C20H42O5)n
CAS Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

non-ionic

Quality Level

mol wt

average Mn ~362

refractive index

n20/D 1.451 (lit.)

density

0.95 g/mL at 25 °C (lit.)

HLB

9

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCO

InChI

1S/C20H42O5/c1-2-3-4-5-6-7-8-9-10-11-13-22-15-17-24-19-20-25-18-16-23-14-12-21/h21H,2-20H2,1H3

InChI key

WPMWEFXCIYCJSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brij® L4 is a non-ionic surfactant. It comprises an alkyl (C12) tail attached to an oxyethylene headgroup.

Application

Brij® L4 has been used:
  • as a surfactant for the synthesis of copper(I) oxide (Cu2O) spherical nanoparticles using a microemulsion technique
  • as a non-ionic surfactant for preparing reverse micelles to synthesize Pt3Co alloy nanoparticles
  • as a surfactant to enhance the efficiency of interfacial coupling of hydrophobic materials and to make hydrogel

Biochem/physiol Actions

Brij® L4 exhibits amphiphilic properties. Brij L4 contains hydrophobic and hydrophilic regions that can form micelles and disperse hydrophobic drugs into the core of micelles. This in turn enhances drug solubility. Brij L4 also elicits a strong inhibitory effect against breast cancer resistance protein (BCRP) mediated drug efflux. It inhibits Taq adsorption better when compared to other commonly used surfactants. Brij L4 supports droplet polymerase chain reaction (PCR) in picoliter reaction volumes at standard Taq concentrations.

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper (i) oxide nanoparticle-mediated C-C couplings for synthesis of polyphenylenediethynylenes: Evidence for a homogeneous catalytic pathway
Pary FF, et al.
Catalysis Science & Technology, 11(7), 2414-2421 (2021)
Synthesis of Pt3Co alloy nanocatalyst via reverse micelle for oxygen reduction reaction in PEMFCs
Liu Z, et al.
Topics in Catalysis, 49(3), 241-250 (2008)
Paul Le Floch et al.
ACS applied materials & interfaces, 9(30), 25542-25552 (2017-07-12)
The emergence of stretchable electronics and its potential integration with textiles have highlighted a challenge: Textiles are wearable and washable, but electronic devices are not. Many stretchable conductors have been developed to enable wearable active textiles, but little has been
Jun Koizumi et al.
AJR. American journal of roentgenology, 199(1), 200-207 (2012-06-27)
Balloon-occluded retrograde transvenous obliteration has been traditionally based on liquid sclerotherapy. However, overdose and systemic spillage of liquid sclerosant can cause severe complications, such as hemolysis, which lead to hemoglobinuria, allergy, acute respiratory distress syndrome, and other disorders. The purpose
Jacomina W van den Esschert et al.
Annals of surgery, 255(2), 311-318 (2012-01-14)
To compare the safety and hypertrophy response after portal vein embolization (PVE) using 2 absorbable and 3 permanent embolization materials. Portal vein embolization is used to increase future remnant liver volume preoperatively. Application of temporary, absorbable embolization materials could be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service